Learn More
Grainger Pipe Marker Nitrogen 5 in H 8 in W
Catalog No. 502752901
Encompass_Preferred
The Brady Pipe Marker Nitrogen is a pre-printed pipe marker that identifies nitrogen gas lines. It is made of durable vinyl with a permanent acrylic adhesive backing for easy application. This pipe marker measures 1.5" H x 7" W and features the word "Nitrogen" printed in black on a yellow background. It meets ANSI/OSHA requirements for pipe labeling and has a temperature range of -40°F to +176°F. The adhesive backing allows it to adhere to PVC, steel, aluminum, and other pipe materials. This versatile marker provides quick, clear identification of nitrogen gas piping.
Catalog No. 50-275-2901 Supplier Grainger Supplier No. 14H934
The Brady Pipe Marker Nitrogen is a pre-printed pipe marker that identifies nitrogen gas lines. It is made of durable vinyl with a permanent acrylic adhesive backing for easy application. This pipe marker measures 1.5" H x 7" W and features the word "Nitrogen" printed in black on a yellow background. It meets ANSI/OSHA requirements for pipe labeling and has a temperature range of -40°F to +176°F. The adhesive backing allows it to adhere to PVC, steel, aluminum, and other pipe materials. This versatile marker provides quick, clear identification of nitrogen gas piping.