Learn More
Grainger Pipe Marker Oxygen 4 in H 24 in W
Catalog No. 502750357
Encompass_Preferred
The Brady Pipe Marker for Oxygen is a self-adhesive vinyl pipe marker designed for identifying oxygen gas lines. It has a bright green background with the word "OXYGEN" printed in black lettering. The marker is 6" tall x 24" long and made from durable vinyl that is weather, UV, chemical, and abrasion resistant. It has an aggressive permanent adhesive that bonds to pipes and most surfaces. This pipe marker meets ANSI/OSHA requirements for labeling oxygen gas piping systems. It provides clear hazard communication and identification for improved safety.
Catalog No. 50-275-0357 Supplier Grainger Supplier No. 14H943
The Brady Pipe Marker for Oxygen is a self-adhesive vinyl pipe marker designed for identifying oxygen gas lines. It has a bright green background with the word "OXYGEN" printed in black lettering. The marker is 6" tall x 24" long and made from durable vinyl that is weather, UV, chemical, and abrasion resistant. It has an aggressive permanent adhesive that bonds to pipes and most surfaces. This pipe marker meets ANSI/OSHA requirements for labeling oxygen gas piping systems. It provides clear hazard communication and identification for improved safety.