Learn More
Grainger Pipe Marker Plant Air 5 in H 8 in W
Catalog No. 502752906
Encompass_Preferred
The Grainger Snap-On Pipe Markers are used to identify pipes and their contents in industrial facilities. They are made of durable vinyl with fade-resistant ink and feature a snap-on design that allows for easy installation and removal. The markers come in a variety of background colors and are offered in sizes ranging from 1/2" to 4" diameter to fit different pipe sizes. They can withstand temperatures from -40°F to 160°F. Overall, these pipe markers from Grainger provide an efficient and customizable way to label pipes for safety and organization.
Catalog No. 50-275-2906 Supplier Grainger Supplier No. 14H949
The Grainger Snap-On Pipe Markers are used to identify pipes and their contents in industrial facilities. They are made of durable vinyl with fade-resistant ink and feature a snap-on design that allows for easy installation and removal. The markers come in a variety of background colors and are offered in sizes ranging from 1/2" to 4" diameter to fit different pipe sizes. They can withstand temperatures from -40°F to 160°F. Overall, these pipe markers from Grainger provide an efficient and customizable way to label pipes for safety and organization.