Learn More
Grainger Pipe Marker Plant Water 26 in H 12 in W
Catalog No. 502750355
Encompass_Preferred
The Brady Pipe Marker for Plant Water is a self-adhesive vinyl pipe marker that identifies plant water pipes. It has a blue background with white text reading "PLANT WATER" and measures 1.5" high x 9" long. This pipe marker is made from flexible, conformable vinyl with a permanent acrylic adhesive that adheres to pipes, valves, and fittings. It is suitable for indoor and outdoor use, resists UV rays and abrasion, and has a temperature range of -40°F to 176°F. The pipe marker meets ANSI standards for pipe marking and OSHA regulations. It comes in a pack of 10 markers.
Catalog No. 50-275-0355 Supplier Grainger Supplier No. 14H952
Add to Cart
Add to Cart
The Brady Pipe Marker for Plant Water is a self-adhesive vinyl pipe marker that identifies plant water pipes. It has a blue background with white text reading "PLANT WATER" and measures 1.5" high x 9" long. This pipe marker is made from flexible, conformable vinyl with a permanent acrylic adhesive that adheres to pipes, valves, and fittings. It is suitable for indoor and outdoor use, resists UV rays and abrasion, and has a temperature range of -40°F to 176°F. The pipe marker meets ANSI standards for pipe marking and OSHA regulations. It comes in a pack of 10 markers.