Learn More
Grainger Pipe Marker Plant Water 5 in H 8 in W
Catalog No. 502751305
Encompass_Preferred
The Grainger Plant Water Pipe Marker is a vinyl label designed to identify pipes carrying water in industrial facilities. It features the words "PLANT WATER" in black text on a blue background. The marker has an aggressive adhesive that adheres to PVC, iron, steel, aluminum, and stainless pipe surfaces. It measures 4 inches wide by 8 inches long and has rounded corners. This durable marker resists chemicals, abrasion, moisture, and fading. It can withstand temperatures from -40°F to 200°F. The Grainger Plant Water Pipe Marker provides clear identification and organization for plant water pipes.
Catalog No. 50-275-1305 Supplier Grainger Supplier No. 14H953
The Grainger Plant Water Pipe Marker is a vinyl label designed to identify pipes carrying water in industrial facilities. It features the words "PLANT WATER" in black text on a blue background. The marker has an aggressive adhesive that adheres to PVC, iron, steel, aluminum, and stainless pipe surfaces. It measures 4 inches wide by 8 inches long and has rounded corners. This durable marker resists chemicals, abrasion, moisture, and fading. It can withstand temperatures from -40°F to 200°F. The Grainger Plant Water Pipe Marker provides clear identification and organization for plant water pipes.