Learn More
Grainger Pipe Marker Plant Water 9 in H 8 in W
Catalog No. 502752916
Encompass_Preferred
The Brady Pipe Marker for Plant Water is a self-adhesive vinyl pipe marker that identifies plant water pipes. It has a blue background with white text that reads "PLANT WATER" in 1-inch tall letters. The marker is 6 inches wide and comes in lengths of 9, 12, 18, and 24 inches to fit various pipe diameters. It is made from flexible, conformable vinyl that resists UV light, abrasion, and chemicals. This durable marker provides quick visual identification of plant water pipes to promote safety and prevent cross-contamination. It meets ANSI standards for pipe marking.
Catalog No. 50-275-2916 Supplier Grainger Supplier No. 14H951
The Brady Pipe Marker for Plant Water is a self-adhesive vinyl pipe marker that identifies plant water pipes. It has a blue background with white text that reads "PLANT WATER" in 1-inch tall letters. The marker is 6 inches wide and comes in lengths of 9, 12, 18, and 24 inches to fit various pipe diameters. It is made from flexible, conformable vinyl that resists UV light, abrasion, and chemicals. This durable marker provides quick visual identification of plant water pipes to promote safety and prevent cross-contamination. It meets ANSI standards for pipe marking.