Learn More
Grainger Pipe Marker Potable Water 26in H 12in W
Catalog No. 502751311
Encompass_Preferred
The Grainger Pipe Marker for Potable Water is a 12" wide x 26" high vinyl marker with adhesive backing that identifies potable water pipes. It has bold black text reading "POTABLE WATER" on a blue background. The vinyl material is fade, tear, and scratch resistant for long-lasting identification. It has an operating temperature range of -40°F to 176°F. This pipe marker meets ANSI/OSHA requirements for pipe labeling and helps identify potable water pipes for safety. It can be applied to pipes, valves, fittings, and equipment.
Catalog No. 50-275-1311 Supplier Grainger Supplier No. 14H955
The Grainger Pipe Marker for Potable Water is a 12" wide x 26" high vinyl marker with adhesive backing that identifies potable water pipes. It has bold black text reading "POTABLE WATER" on a blue background. The vinyl material is fade, tear, and scratch resistant for long-lasting identification. It has an operating temperature range of -40°F to 176°F. This pipe marker meets ANSI/OSHA requirements for pipe labeling and helps identify potable water pipes for safety. It can be applied to pipes, valves, fittings, and equipment.