Learn More
Grainger Pipe Marker Process Water 5 in H 8 in W
Catalog No. 502751438
Encompass_Preferred
The Grainger Approved Pipe Marker is a self-adhesive vinyl pipe marker that can be used for identifying pipes, valves, fittings, and equipment. It has a high-tack adhesive backing and is made of durable vinyl that resists fading, abrasion, moisture, and chemicals. This pipe marker comes in a variety of background colors and can be customized with text indicating the pipe contents and flow direction. It meets ANSI/OSHA requirements for pipe labeling. The pipe marker measures 9" x 3" and is sold individually. It can withstand temperatures up to 176°F.
Catalog No. 50-275-1438 Supplier Grainger Supplier No. 14H964
The Grainger Approved Pipe Marker is a self-adhesive vinyl pipe marker that can be used for identifying pipes, valves, fittings, and equipment. It has a high-tack adhesive backing and is made of durable vinyl that resists fading, abrasion, moisture, and chemicals. This pipe marker comes in a variety of background colors and can be customized with text indicating the pipe contents and flow direction. It meets ANSI/OSHA requirements for pipe labeling. The pipe marker measures 9" x 3" and is sold individually. It can withstand temperatures up to 176°F.