Learn More
Grainger Pipe Marker Process Water 9 in H 8 in W
Catalog No. 502752917
Encompass_Preferred
The Brady Pipe Marker for Process Water is a self-adhesive vinyl pipe marker that identifies process water pipes. It has a blue background with white text reading "PROCESS WATER" and has an operating temperature range of -40°F to 176°F. This pipe marker meets ANSI/ASME A13.1 standards and has a 5 year outdoor durability. It comes in various sizes to fit pipe diameters from 0.5" to 36".
Catalog No. 50-275-2917 Supplier Grainger Supplier No. 14H962
The Brady Pipe Marker for Process Water is a self-adhesive vinyl pipe marker that identifies process water pipes. It has a blue background with white text reading "PROCESS WATER" and has an operating temperature range of -40°F to 176°F. This pipe marker meets ANSI/ASME A13.1 standards and has a 5 year outdoor durability. It comes in various sizes to fit pipe diameters from 0.5" to 36".