Learn More
Grainger Pipe Marker Steam 26 in H 12 in W
Catalog No. 502750432
Encompass_Preferred
The Brady Pipe Marker for Steam is a self-adhesive vinyl pipe marker that identifies steam pipes. It has a white background with black text reading "STEAM" and directional arrows. The marker is made of flexible, conformable vinyl that resists UV light, abrasion, and chemicals. It has an operating temperature range of -40°F to 176°F. The marker comes in 12" length x 3" width and is sold individually. It meets ANSI/OSHA requirements for pipe marking and labeling. The adhesive backing allows it to stick to pipes up to 2" diameter. This pipe marker offers an easy way to identify steam pipes for safety purposes.
Catalog No. 50-275-0432 Supplier Grainger Supplier No. 14H986
The Brady Pipe Marker for Steam is a self-adhesive vinyl pipe marker that identifies steam pipes. It has a white background with black text reading "STEAM" and directional arrows. The marker is made of flexible, conformable vinyl that resists UV light, abrasion, and chemicals. It has an operating temperature range of -40°F to 176°F. The marker comes in 12" length x 3" width and is sold individually. It meets ANSI/OSHA requirements for pipe marking and labeling. The adhesive backing allows it to stick to pipes up to 2" diameter. This pipe marker offers an easy way to identify steam pipes for safety purposes.