Learn More
Grainger Pipe Marker Steam 5 in H 8 in W
Catalog No. 502750433
Encompass_Preferred
The Brady 14H987 Pipe Marker for Steam is a self-adhesive vinyl pipe marker that identifies steam pipes. It has a white background with black text reading "STEAM" in 1-1/2 inch tall letters. The marker is 6 inches wide and comes in a 36 inch length roll. It is made from flexible, conformable vinyl that resists UV light and abrasion. The adhesive backing allows it to stick to pipes up to 1-3/4 inches in diameter. This steam pipe marker meets ANSI/OSHA requirements for pipe labeling and identification. It provides clear visibility to identify steam lines for safety purposes.
Catalog No. 50-275-0433 Supplier Grainger Supplier No. 14H987
The Brady 14H987 Pipe Marker for Steam is a self-adhesive vinyl pipe marker that identifies steam pipes. It has a white background with black text reading "STEAM" in 1-1/2 inch tall letters. The marker is 6 inches wide and comes in a 36 inch length roll. It is made from flexible, conformable vinyl that resists UV light and abrasion. The adhesive backing allows it to stick to pipes up to 1-3/4 inches in diameter. This steam pipe marker meets ANSI/OSHA requirements for pipe labeling and identification. It provides clear visibility to identify steam lines for safety purposes.