Learn More
Grainger Pipe Marker Storm Drain 5 in H 8 in W
Catalog No. 502751061
Encompass_Preferred
The Brady Pipe Marker for Storm Drains is a pre-printed, self-adhesive vinyl label designed to identify storm drains and warn against dumping pollutants. This marker has a legend that reads "Dump No Waste - Drains to Stream" and has an image of a fish. It is made of durable vinyl that is UV, water, and chemical resistant. The marker measures 4" W x 6" H and has an aggressive permanent adhesive that adheres to PVC, concrete, metal, and other low-energy surfaces. It meets ANSI Z535.1 standards for safety labels. This product helps facilities comply with stormwater regulations and prevents pollutants from entering waterways.
Catalog No. 50-275-1061 Supplier Grainger Supplier No. 14H991
The Brady Pipe Marker for Storm Drains is a pre-printed, self-adhesive vinyl label designed to identify storm drains and warn against dumping pollutants. This marker has a legend that reads "Dump No Waste - Drains to Stream" and has an image of a fish. It is made of durable vinyl that is UV, water, and chemical resistant. The marker measures 4" W x 6" H and has an aggressive permanent adhesive that adheres to PVC, concrete, metal, and other low-energy surfaces. It meets ANSI Z535.1 standards for safety labels. This product helps facilities comply with stormwater regulations and prevents pollutants from entering waterways.