Learn More
Grainger Pipe Marker Vacuum 9 in H 8 in W
Catalog No. 502751082
Encompass_Preferred
The Brady Pipe Marker Vacuum is a handheld device used to apply pre-printed self-adhesive markers onto pipes, valves, and other surfaces. It uses a vacuum to hold the marker in place for quick, easy application. This model has a compact pistol grip design and requires no batteries or electricity. It can apply markers up to 5" wide. The vacuum pad is made of rubber for durability. This is a useful tool for efficiently labeling pipes during installation or maintenance.
Catalog No. 50-275-1082 Supplier Grainger Supplier No. 14J002
The Brady Pipe Marker Vacuum is a handheld device used to apply pre-printed self-adhesive markers onto pipes, valves, and other surfaces. It uses a vacuum to hold the marker in place for quick, easy application. This model has a compact pistol grip design and requires no batteries or electricity. It can apply markers up to 5" wide. The vacuum pad is made of rubber for durability. This is a useful tool for efficiently labeling pipes during installation or maintenance.