Learn More
Grainger Pipe Marker Vent 5 in H 8 in W
Catalog No. 502751059
Encompass_Preferred
The Brady 14J010 Pipe Marker Vent is a self-adhesive vinyl pipe marker that identifies vent pipes. It has a white background with black text reading "VENT" in 1-1/2 inch tall letters. The marker is 6 inches wide and comes in a pack of 10. It is made from flexible, conformable vinyl that adheres to pipes up to 1-1/4 inches in diameter. The adhesive is resistant to UV rays, water, chemicals, abrasion, and temperatures up to 176°F. This versatile marker offers an easy way to clearly identify vent pipes for safety and compliance.
Catalog No. 50-275-1059 Supplier Grainger Supplier No. 14J010
The Brady 14J010 Pipe Marker Vent is a self-adhesive vinyl pipe marker that identifies vent pipes. It has a white background with black text reading "VENT" in 1-1/2 inch tall letters. The marker is 6 inches wide and comes in a pack of 10. It is made from flexible, conformable vinyl that adheres to pipes up to 1-1/4 inches in diameter. The adhesive is resistant to UV rays, water, chemicals, abrasion, and temperatures up to 176°F. This versatile marker offers an easy way to clearly identify vent pipes for safety and compliance.