Learn More
Grainger Pipe Marker Vent 5 in H 8 in W
Catalog No. 502750435
Encompass_Preferred
The Brady Pipe Marker Vent 14J007 is a self-adhesive vinyl pipe marker designed for identifying vent pipes. It features a high-tack adhesive that adheres to PVC, ABS, fiberglass, and metal pipes from -40°F to 200°F. This 3" x 6" marker has bold black text reading "VENT" on a yellow background. It is made from flexible, fade-resistant vinyl that resists moisture, abrasion, and chemicals. The adhesive backing allows for quick and easy application without tools or fasteners. This versatile marker provides an easy way to label vent pipes for identification and safety purposes.
Catalog No. 50-275-0435 Supplier Grainger Supplier No. 14J007
The Brady Pipe Marker Vent 14J007 is a self-adhesive vinyl pipe marker designed for identifying vent pipes. It features a high-tack adhesive that adheres to PVC, ABS, fiberglass, and metal pipes from -40°F to 200°F. This 3" x 6" marker has bold black text reading "VENT" on a yellow background. It is made from flexible, fade-resistant vinyl that resists moisture, abrasion, and chemicals. The adhesive backing allows for quick and easy application without tools or fasteners. This versatile marker provides an easy way to label vent pipes for identification and safety purposes.