Learn More
Grainger Pipe Marker Vent 9 in H 8 in W
Catalog No. 502751134
Encompass_Preferred
The Brady Pipe Marker Vent 14J008 is a self-adhesive vinyl pipe marker designed for identifying vent pipes. It has a legend indicating "Vent" and is colored yellow for quick visual identification. The marker is made from flexible vinyl with a permanent acrylic adhesive that adheres to pipes. It measures 1-1/2" H x 6" W. This pipe marker meets ANSI/OSHA requirements for pipe labeling and identification. It provides a neat, professional look and is UV, chemical, oil, and scratch resistant. The Brady Pipe Marker Vent 14J008 is suitable for indoor and outdoor use and can withstand temperatures from -40°F to 176°F when applied properly.
Catalog No. 50-275-1134 Supplier Grainger Supplier No. 14J008
The Brady Pipe Marker Vent 14J008 is a self-adhesive vinyl pipe marker designed for identifying vent pipes. It has a legend indicating "Vent" and is colored yellow for quick visual identification. The marker is made from flexible vinyl with a permanent acrylic adhesive that adheres to pipes. It measures 1-1/2" H x 6" W. This pipe marker meets ANSI/OSHA requirements for pipe labeling and identification. It provides a neat, professional look and is UV, chemical, oil, and scratch resistant. The Brady Pipe Marker Vent 14J008 is suitable for indoor and outdoor use and can withstand temperatures from -40°F to 176°F when applied properly.