Learn More
Grainger Pipe Marker Water 4 in H 24 in W
Catalog No. 502752908
Encompass_Preferred
The Brady Pipe Marker for Water is a self-adhesive vinyl pipe marker that identifies water pipes. It has a blue background with the word "WATER" in large white text for high visibility. The marker is 6" wide x 9" tall and made from durable vinyl that is weather and UV resistant. It has an aggressive adhesive that bonds to PVC, steel, and other common pipe materials. This pipe marker meets ANSI/OSHA requirements for pipe labeling and identification. It provides an easy and organized way to label water pipes for safety and compliance.
Catalog No. 50-275-2908 Supplier Grainger Supplier No. 14J022
The Brady Pipe Marker for Water is a self-adhesive vinyl pipe marker that identifies water pipes. It has a blue background with the word "WATER" in large white text for high visibility. The marker is 6" wide x 9" tall and made from durable vinyl that is weather and UV resistant. It has an aggressive adhesive that bonds to PVC, steel, and other common pipe materials. This pipe marker meets ANSI/OSHA requirements for pipe labeling and identification. It provides an easy and organized way to label water pipes for safety and compliance.