Learn More
Grainger Pipe Marker Water 9 in H 8 in W
Catalog No. 502750522
Encompass_Preferred
The Brady Pipe Marker for water is a self-adhesive vinyl pipe marker that identifies water pipes. It has a blue background with white text reading "WATER" and has an aggressive adhesive that adheres to PVC, ABS, fiberglass, and metal pipes. The marker measures 1.5 inches wide by 9 inches long. It is made in the USA and meets ANSI standards for pipe marking. The marker has a temperature range of -40°F to 176°F and is UV, chemical, oil, and grease resistant. It has an outdoor durability of 5 years.
Catalog No. 50-275-0522 Supplier Grainger Supplier No. 14J019
The Brady Pipe Marker for water is a self-adhesive vinyl pipe marker that identifies water pipes. It has a blue background with white text reading "WATER" and has an aggressive adhesive that adheres to PVC, ABS, fiberglass, and metal pipes. The marker measures 1.5 inches wide by 9 inches long. It is made in the USA and meets ANSI standards for pipe marking. The marker has a temperature range of -40°F to 176°F and is UV, chemical, oil, and grease resistant. It has an outdoor durability of 5 years.