Learn More
Grainger Pipe Markr Boiler Blowdown 26in H 12in W
Catalog No. 502749303
Encompass_Preferred
The Brady Pipe Marker for Boiler Feed Water is a self-adhesive vinyl pipe marker that identifies boiler feed water pipes. It has a white background with black text that reads "BOILER FEED WATER" in 1-1/2 inch tall letters. This pipe marker meets ANSI standards for pipe marking and has a temperature range of -40°F to 176°F. It has an outdoor durability of 5 years. The pipe marker comes in rolls that are 1-1/2 inches wide x 50 feet long. It can be applied on pipes up to 2 inches in diameter. This product helps identify boiler feed water pipes for safety and compliance.
Catalog No. 50-274-9303 Supplier Grainger Supplier No. 14H763
The Brady Pipe Marker for Boiler Feed Water is a self-adhesive vinyl pipe marker that identifies boiler feed water pipes. It has a white background with black text that reads "BOILER FEED WATER" in 1-1/2 inch tall letters. This pipe marker meets ANSI standards for pipe marking and has a temperature range of -40°F to 176°F. It has an outdoor durability of 5 years. The pipe marker comes in rolls that are 1-1/2 inches wide x 50 feet long. It can be applied on pipes up to 2 inches in diameter. This product helps identify boiler feed water pipes for safety and compliance.