Learn More
Grainger Pipe Markr Domestic Cold Water 4 1/4in H
Catalog No. 502753245
Encompass_Preferred
The Brady Pipe Marker - Domestic Cold Water is a self-adhesive vinyl pipe marker that identifies cold domestic water pipes. It has a blue background with white text that reads "Domestic Cold Water". The marker measures 1-1/2" wide x 9" long and is made from 6 mil vinyl. It can withstand temperatures from -40°F to 176°F. This pipe marker meets ANSI/OSHA requirements for pipe labeling and helps identify pipes for safety purposes. It comes in a pack of 10 markers.
Catalog No. 50-275-3245 Supplier Grainger Supplier No. 14H824
The Brady Pipe Marker - Domestic Cold Water is a self-adhesive vinyl pipe marker that identifies cold domestic water pipes. It has a blue background with white text that reads "Domestic Cold Water". The marker measures 1-1/2" wide x 9" long and is made from 6 mil vinyl. It can withstand temperatures from -40°F to 176°F. This pipe marker meets ANSI/OSHA requirements for pipe labeling and helps identify pipes for safety purposes. It comes in a pack of 10 markers.