Learn More
Grainger Pipe Mrkr Chilld Water Spply 5in H 8in W
Catalog No. 502751847
Encompass_Preferred
The Brady Pipe Marker for Chilled Water is a self-adhesive vinyl pipe marker that identifies pipes carrying chilled water. It has a blue background with white text reading "CHILLED WATER" and has an operating temperature range of -40°F to 200°F. The marker measures 1.5" height x 9" width and has an adhesive backing for easy application. It is made from flexible, conformable vinyl that resists UV light, abrasion, and chemicals. This pipe marker from Brady helps identify chilled water pipes for safety and compliance.
Catalog No. 50-275-1847 Supplier Grainger Supplier No. 14H787
The Brady Pipe Marker for Chilled Water is a self-adhesive vinyl pipe marker that identifies pipes carrying chilled water. It has a blue background with white text reading "CHILLED WATER" and has an operating temperature range of -40°F to 200°F. The marker measures 1.5" height x 9" width and has an adhesive backing for easy application. It is made from flexible, conformable vinyl that resists UV light, abrasion, and chemicals. This pipe marker from Brady helps identify chilled water pipes for safety and compliance.