Learn More
Grainger Pipe Mrkr Domestic Cold Watr 9in H 8in W
Catalog No. 502749061
Encompass_Preferred
The Brady Pipe Marker for Domestic Cold Water is a self-adhesive vinyl pipe marker that identifies cold domestic water pipes. It has a blue background with white text reading "Domestic Cold Water" and has an operating temperature range of -40°F to 176°F. This pipe marker meets ANSI/OSHA requirements for pipe labeling and has a 5 year outdoor durability. It comes in various sizes to fit pipe diameters from 0.5" to 36".
Catalog No. 50-274-9061 Supplier Grainger Supplier No. 14H825
The Brady Pipe Marker for Domestic Cold Water is a self-adhesive vinyl pipe marker that identifies cold domestic water pipes. It has a blue background with white text reading "Domestic Cold Water" and has an operating temperature range of -40°F to 176°F. This pipe marker meets ANSI/OSHA requirements for pipe labeling and has a 5 year outdoor durability. It comes in various sizes to fit pipe diameters from 0.5" to 36".