Learn More
Grainger Pipe Mrkr Domestic Hot Watr Return 5in H
Catalog No. 502749025
Encompass_Preferred
The Brady Pipe Marker for Domestic Hot Water is a self-adhesive vinyl pipe marker that identifies hot water pipes. It has a blue background with white text that reads "Domestic Hot Water" and has an operating temperature range of -40°F to 176°F. The marker measures 1.5" W x 9" L and has an adhesive backing for easy application. It is made from flexible, conformable vinyl that resists UV light, abrasion, and chemicals. This pipe marker from Brady helps identify hot water pipes for safety purposes and meets ANSI A13.1 and NFPA 99 standards.
Catalog No. 50-274-9025 Supplier Grainger Supplier No. 14H835
The Brady Pipe Marker for Domestic Hot Water is a self-adhesive vinyl pipe marker that identifies hot water pipes. It has a blue background with white text that reads "Domestic Hot Water" and has an operating temperature range of -40°F to 176°F. The marker measures 1.5" W x 9" L and has an adhesive backing for easy application. It is made from flexible, conformable vinyl that resists UV light, abrasion, and chemicals. This pipe marker from Brady helps identify hot water pipes for safety purposes and meets ANSI A13.1 and NFPA 99 standards.