Learn More
Grainger Pipe Mrkr Domstc Cold Watr 26in H 12in W
Catalog No. 502753246
Encompass_Preferred
The Brady Pipe Marker for Domestic Cold Water is a self-adhesive vinyl pipe marker that identifies cold domestic water pipes. It has a blue background with white text reading "Domestic Cold Water" and has an operating temperature range of -40°F to 176°F. This pipe marker meets ANSI standards for pipe marking and identification. It comes in rolls of various lengths and widths to accommodate different pipe diameters. This product provides clear visual identification of cold domestic water pipes to promote safety and prevent accidental misconnections. The vinyl material is durable for long-lasting pipe identification.
Catalog No. 50-275-3246 Supplier Grainger Supplier No. 14H826
The Brady Pipe Marker for Domestic Cold Water is a self-adhesive vinyl pipe marker that identifies cold domestic water pipes. It has a blue background with white text reading "Domestic Cold Water" and has an operating temperature range of -40°F to 176°F. This pipe marker meets ANSI standards for pipe marking and identification. It comes in rolls of various lengths and widths to accommodate different pipe diameters. This product provides clear visual identification of cold domestic water pipes to promote safety and prevent accidental misconnections. The vinyl material is durable for long-lasting pipe identification.