Learn More
Grainger Pipe Mrkr Fire Prtction Watr 5in H 8in W
Catalog No. 502750017
Encompass_Preferred
The Grainger 14H847 is a 14-inch diameter, high-speed steel hole saw designed for cutting holes in wood, plastic, and light metals. It features bi-metal construction with M42 high-speed steel teeth brazed to a carbon steel body for durability and long life. The 14H847 has a 1-1/8-inch arbor hole and comes with a pilot bit for guiding the hole saw. Recommended operating speed is up to 2500 RPM with proper lubrication. Overall length is 4-1/2 inches.
Catalog No. 50-275-0017 Supplier Grainger Supplier No. 14H847
The Grainger 14H847 is a 14-inch diameter, high-speed steel hole saw designed for cutting holes in wood, plastic, and light metals. It features bi-metal construction with M42 high-speed steel teeth brazed to a carbon steel body for durability and long life. The 14H847 has a 1-1/8-inch arbor hole and comes with a pilot bit for guiding the hole saw. Recommended operating speed is up to 2500 RPM with proper lubrication. Overall length is 4-1/2 inches.