Learn More
Grainger Pipe Mrkr Heating Watr Retrn 9in H 8in W
Catalog No. 502750430
Encompass_Preferred
The Brady Pipe Marker for Heating Water Return is a self-adhesive vinyl pipe marker that identifies heating water return pipes. It has a legend that reads "HEATING WATER RETURN" in black text on a yellow background. The marker is 6" tall x 18" long and made from flexible, conformable vinyl that resists chemicals, abrasion, and moisture. It has an aggressive adhesive that adheres well to pipes. This pipe marker meets ANSI A13.1 and NFPA 99 standards for pipe labeling and helps identify pipes for safety and maintenance. It comes in a pack of 10 markers.
Catalog No. 50-275-0430 Supplier Grainger Supplier No. 14H880
The Brady Pipe Marker for Heating Water Return is a self-adhesive vinyl pipe marker that identifies heating water return pipes. It has a legend that reads "HEATING WATER RETURN" in black text on a yellow background. The marker is 6" tall x 18" long and made from flexible, conformable vinyl that resists chemicals, abrasion, and moisture. It has an aggressive adhesive that adheres well to pipes. This pipe marker meets ANSI A13.1 and NFPA 99 standards for pipe labeling and helps identify pipes for safety and maintenance. It comes in a pack of 10 markers.