Learn More
Grainger Pipe Mrkr Heating Watr Spply 5in H 8in W
Catalog No. 502751094
Encompass_Preferred
The Brady Pipe Marker for Heating Water Supply is a self-adhesive vinyl pipe marker that identifies heating water supply pipes. It has a white background with black text that reads "HEATING WATER SUPPLY" and has directional flow arrows. The marker is made of flexible, conformable vinyl that adheres to pipes up to 2" in diameter. It meets ANSI A13.1 and NFPA 99 standards for pipe marking and has a temperature range of -40°F to 200°F. The marker measures 9" x 3" and comes in a pack of 10. It provides clear identification and organization for heating water supply pipes.
Catalog No. 50-275-1094 Supplier Grainger Supplier No. 14H886
The Brady Pipe Marker for Heating Water Supply is a self-adhesive vinyl pipe marker that identifies heating water supply pipes. It has a white background with black text that reads "HEATING WATER SUPPLY" and has directional flow arrows. The marker is made of flexible, conformable vinyl that adheres to pipes up to 2" in diameter. It meets ANSI A13.1 and NFPA 99 standards for pipe marking and has a temperature range of -40°F to 200°F. The marker measures 9" x 3" and comes in a pack of 10. It provides clear identification and organization for heating water supply pipes.