Learn More
Grainger Pipe Mrkr High Prssr Steam 26in H 12in W
Catalog No. 502751067
Encompass_Preferred
The Brady Pipe Marker High Pressure Steam 14H889 is a self-adhesive vinyl pipe marker that identifies high pressure steam pipes. It has a high performance acrylic adhesive and is made of durable vinyl that resists UV light, chemicals, abrasion, and moisture. This pipe marker has an operating temperature range of -40°F to 200°F and conforms to ANSI A13.1 and NFPA 99 standards. The marker comes in yellow with black text reading "HIGH PRESSURE STEAM" and is available in various sizes to fit pipe diameters from 0.5 to 6 inches. This product provides clear identification and warning for high pressure steam pipes.
Catalog No. 50-275-1067 Supplier Grainger Supplier No. 14H889
The Brady Pipe Marker High Pressure Steam 14H889 is a self-adhesive vinyl pipe marker that identifies high pressure steam pipes. It has a high performance acrylic adhesive and is made of durable vinyl that resists UV light, chemicals, abrasion, and moisture. This pipe marker has an operating temperature range of -40°F to 200°F and conforms to ANSI A13.1 and NFPA 99 standards. The marker comes in yellow with black text reading "HIGH PRESSURE STEAM" and is available in various sizes to fit pipe diameters from 0.5 to 6 inches. This product provides clear identification and warning for high pressure steam pipes.