Learn More
Grainger Pipe Mrkr Sdium Hypochlorite 9in H 8in W
Catalog No. 502750015
Encompass_Preferred
The Grainger 14H977 is a 3M Speedglas Welding Helmet that features auto-darkening technology to protect the eyes and face from sparks, spatter, and harmful UV/IR rays while welding or cutting. This helmet has a large viewing area for increased visibility. It has adjustable sensitivity and delay controls so you can customize settings for optimal comfort and protection. The helmet operates on solar cells so no batteries are required. It meets ANSI Z87.1 standards for impact protection. The Speedglas helmet provides reliable eye safety and optical clarity for welders.
Catalog No. 50-275-0015 Supplier Grainger Supplier No. 14H977
The Grainger 14H977 is a 3M Speedglas Welding Helmet that features auto-darkening technology to protect the eyes and face from sparks, spatter, and harmful UV/IR rays while welding or cutting. This helmet has a large viewing area for increased visibility. It has adjustable sensitivity and delay controls so you can customize settings for optimal comfort and protection. The helmet operates on solar cells so no batteries are required. It meets ANSI Z87.1 standards for impact protection. The Speedglas helmet provides reliable eye safety and optical clarity for welders.