Learn More
MSC Vermont Gage Plug Gage, Class X, Tool Steel, 0.0044 in., 1 ea.
Catalog No. 5000511273
Encompass
The Class X Go (Plus) plug gage serves as a reliable measurement standard for calibrating inspection equipment or direct part gaging in shop floor and metrology laboratory environments. This 0.0044 in. tool is engineered for precision and longevity, ensuring accurate quality control.
  • Constructed from durable tool steel for extended service life
  • Features a 4 microinch finish or better
  • Hardness rated at 60/65 HRC
  • Manufactured with Class X tolerance of +.00004 in./-.00000 in.
  • Round within 1/2 of the total tolerance
  • Certificate of accuracy included with every unit
  • Made in U.S.A.
Catalog No. 50-005-11273 Supplier MSC Supplier No. 05450044
The Class X Go (Plus) plug gage serves as a reliable measurement standard for calibrating inspection equipment or direct part gaging in shop floor and metrology laboratory environments. This 0.0044 in. tool is engineered for precision and longevity, ensuring accurate quality control.
  • Constructed from durable tool steel for extended service life
  • Features a 4 microinch finish or better
  • Hardness rated at 60/65 HRC
  • Manufactured with Class X tolerance of +.00004 in./-.00000 in.
  • Round within 1/2 of the total tolerance
  • Certificate of accuracy included with every unit
  • Made in U.S.A.