Learn More
MSC Balax Pipe Thread Plug Gage, 1/4-18 NPT, Tapered, Single End, 1 ea
Catalog No. 5000516833
Encompass
This tapered pipe thread plug gage is designed for precise inspection of NPT threads. Engineered for accuracy and durability, it features a single-end configuration for efficient thread verification in industrial and metalworking applications.
  • Constructed from high speed tool steel for enhanced wear resistance
  • Hardness rating of 63 to 65 Rockwell
  • Bright finish for improved visibility
  • Equipped with a #3 Taperlock handle
  • Overall length is 1.45 in. with 0.45 in. thread length
  • Designed for 1/4-18 NPT L1 thread classification
Catalog No. 50-005-16833 Supplier MSC Supplier No. 05450036
This tapered pipe thread plug gage is designed for precise inspection of NPT threads. Engineered for accuracy and durability, it features a single-end configuration for efficient thread verification in industrial and metalworking applications.
  • Constructed from high speed tool steel for enhanced wear resistance
  • Hardness rating of 63 to 65 Rockwell
  • Bright finish for improved visibility
  • Equipped with a #3 Taperlock handle
  • Overall length is 1.45 in. with 0.45 in. thread length
  • Designed for 1/4-18 NPT L1 thread classification