Learn More
Waters Corp Pipe Thread Reducing Adapter, Brass
Supplier: Waters Corp WAT050224
This pipe thread reducing adapter is made of brass and is designed for use with various liquid chromatography (LC) systems.
- Made of brass
- Compatible with LC Module 1
- Compatible with 600 MultiSolvent Delivery System
- Compatible with 610 PowerLine Isocratic Pump
- Compatible with 626 LC System
- Designed for liquid chromatography systems