Learn More
Medchemexpress LLC HY-Y0669 100mg Medchemexpress, Pipecolic acid CAS:535-75-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYY0669100MG
Encompass_Preferred
Medchemexpress, HY-Y0669 100mg Pipecolic acid CAS:535-75-1 Pipecolic acid is a metabolite of lysine. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0764