Learn More
Medchemexpress LLC Pipecolic acid | 535-75-1 | MFCD00064347 | 99.9% | 129.16 | C6H11NO2 | 50g
SDP
Catalog No. 5000268976
Encompass_Preferred
Pipecolic acid is a metabolite of lysine and an important precursor for many useful microbial secondary metabolites Pipecolic acid slows ferroptosis in diabetic retinopathy by inhibiting the YAP-GPX4 signaling pathway In addition Pipecolic acid can be used as a diagnostic marker for pyridoxine-dependent epilepsy[1][2][3]
Catalog No. 50-002-68976 Supplier Medchemexpress LLC Supplier No. HYY066950G
Add to Cart
Add to Cart
Pipecolic acid is a metabolite of lysine and an important precursor for many useful microbial secondary metabolites Pipecolic acid slows ferroptosis in diabetic retinopathy by inhibiting the YAP-GPX4 signaling pathway In addition Pipecolic acid can be used as a diagnostic marker for pyridoxine-dependent epilepsy[1][2][3]