Learn More
Medchemexpress LLC Pipecolic acid-d9 | 790612-94-1 | 99.9% | 138.21 | C6H2D9NO2 | 5mg
SDP
Catalog No. 5000269085
Encompass_Preferred
Pipecolic acid-d9 is the deuterium labeled Pipecolic acid Pipecolic acid is a metabolite of lysine and an important precursor for many useful microbial secondary metabolites Pipecolic acid slows ferroptosis in diabetic retinopathy by inhibiting the YAP-GPX4 signaling pathway In addition Pipecolic acid can be used as a diagnostic marker for pyridoxine-dependent epilepsy[1][2][3]
Catalog No. 50-002-69085 Supplier Medchemexpress LLC Supplier No. HYY0669S5MG
Pipecolic acid-d9 is the deuterium labeled Pipecolic acid Pipecolic acid is a metabolite of lysine and an important precursor for many useful microbial secondary metabolites Pipecolic acid slows ferroptosis in diabetic retinopathy by inhibiting the YAP-GPX4 signaling pathway In addition Pipecolic acid can be used as a diagnostic marker for pyridoxine-dependent epilepsy[1][2][3]