Learn More
Medchemexpress LLC Pipecolic acid-d9 | 790612-94-1 | MFCD00064347 | 99.9% | 138.21 g/mol | C6H2D9NO2 | 1mg
SDP
Catalog No. 5000268900
Encompass_Preferred
Pipecolic acid-d9 is the deuterium labeled Pipecolic acid Pipecolic acid is a metabolite of lysine and an important precursor for many useful microbial secondary metabolites Pipecolic acid slows ferroptosis in diabetic retinopathy by inhibiting the YAP-GPX4 signaling pathway In addition Pipecolic acid can be used as a diagnostic marker for pyridoxine-dependent epilepsy[1][2][3]
Catalog No. 50-002-68900 Supplier Medchemexpress LLC Supplier No. HYY0669S1MG
Pipecolic acid-d9 is the deuterium labeled Pipecolic acid Pipecolic acid is a metabolite of lysine and an important precursor for many useful microbial secondary metabolites Pipecolic acid slows ferroptosis in diabetic retinopathy by inhibiting the YAP-GPX4 signaling pathway In addition Pipecolic acid can be used as a diagnostic marker for pyridoxine-dependent epilepsy[1][2][3]