Learn More
TARGETMOL CHEMICALS INC Pipecuronium bromide 10MG
SDP
Catalog No. 502879855
Encompass_Preferred
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Pipecuronium bromide (RGH-1106) is a selective and potent nAChR antagonist that also acts as a non-depolarising steroidal neuromuscular blocker, causing muscle relaxation, often in conjunction with antibiotics. Purity 100%
Catalog No. 50-287-9855 Supplier TARGETMOL CHEMICALS INC Supplier No. T1248110MG
Also available in 1 mg, 5 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Pipecuronium bromide (RGH-1106) is a selective and potent nAChR antagonist that also acts as a non-depolarising steroidal neuromuscular blocker, causing muscle relaxation, often in conjunction with antibiotics. Purity 100%