Learn More
Medchemexpress LLC Pipecuronium bromide | 52212-02-9 | 99.88% | 50 MG
SDP
Catalog No. 5000409394
Encompass_Preferred
Pipecuronium bromide is a potent, long-acting, nondepolarizing steroidal neuromuscular blocking agent (NMBA) and a bisquaternary ammonium compound. It acts as a powerful competitive nAChR antagonist. It does not liberate histamine and has no cardiovascular side effects.
  • Potent, long-acting nondepolarizing steroidal neuromuscular blocking agent
  • Bisquaternary ammonium compound
  • Acts as a powerful competitive nAChR antagonist with a Kd of 3.06 μM
  • Does not liberate histamine
  • No cardiovascular side effects even at doses of 3x ED95
  • Anaphylaxis does not appear to be a problem
  • Sugammadex has a high affinity for Pipecuronium bromide
  • Fewer molecules required to achieve comparable blockade than with Rocuronium
  • Carboxymethylated γ-cyclodextrin can efficiently and completely reverse Pipecuronium bromide-induced neuromuscular block
Catalog No. 50-004-09394 Supplier Medchemexpress LLC Supplier No. HYB0743A50MG
Pipecuronium bromide is a potent, long-acting, nondepolarizing steroidal neuromuscular blocking agent (NMBA) and a bisquaternary ammonium compound. It acts as a powerful competitive nAChR antagonist. It does not liberate histamine and has no cardiovascular side effects.
  • Potent, long-acting nondepolarizing steroidal neuromuscular blocking agent
  • Bisquaternary ammonium compound
  • Acts as a powerful competitive nAChR antagonist with a Kd of 3.06 μM
  • Does not liberate histamine
  • No cardiovascular side effects even at doses of 3x ED95
  • Anaphylaxis does not appear to be a problem
  • Sugammadex has a high affinity for Pipecuronium bromide
  • Fewer molecules required to achieve comparable blockade than with Rocuronium
  • Carboxymethylated γ-cyclodextrin can efficiently and completely reverse Pipecuronium bromide-induced neuromuscular block