Learn More
Medchemexpress LLC Pipecuronium (bromide) | 52212-02-9 | 99.9% | 1 ML
SDP
Catalog No. 5000407088
Encompass_Preferred
Pipecuronium bromide is a potent, long-acting nondepolarizing steroidal neuromuscular blocking agent (NMBA) and a bisquaternary ammonium compound. It acts as a powerful competitive nAChR antagonist with a Kd of 3.06 μM.
  • Potent long-acting neuromuscular blocking agent.
  • Acts as a powerful competitive nAChR antagonist.
  • Approximately 6 to 7 times more potent than Rocuronium.
  • Does not liberate histamine and has no cardiovascular side effects.
  • Neuromuscular block can be reversed by carboxymethylated γ-cyclodextrin.
Catalog No. 50-004-07088 Supplier Medchemexpress LLC Supplier No. HYB0743A10MM/1ML
Pipecuronium bromide is a potent, long-acting nondepolarizing steroidal neuromuscular blocking agent (NMBA) and a bisquaternary ammonium compound. It acts as a powerful competitive nAChR antagonist with a Kd of 3.06 μM.
  • Potent long-acting neuromuscular blocking agent.
  • Acts as a powerful competitive nAChR antagonist.
  • Approximately 6 to 7 times more potent than Rocuronium.
  • Does not liberate histamine and has no cardiovascular side effects.
  • Neuromuscular block can be reversed by carboxymethylated γ-cyclodextrin.