Learn More
Medchemexpress LLC Pipecuronium bromide | 52212-02-9 | MFCD00867696 | 99.9% | 762.70 | C35H62Br2N4O4 | 10 MG
SDP
Catalog No. 5000297375
Encompass_Preferred
Pipecuronium bromide is a long-acting, nondepolarizing steroidal neuromuscular blocking agent supplied as a solid for research use. It functions as a nicotinic acetylcholine receptor antagonist to produce controlled muscle relaxation, and is commonly used in pharmacology and physiology studies requiring reproducible neuromuscular blockade.
  • Long-acting nondepolarizing neuromuscular blocker suitable for research.
  • High reported purity (≈99.9%) for reproducible experimental results.
  • Solid form enables stable storage and accurate dosing.
  • Available in small mass quantities compatible with laboratory assays.
  • Documented storage conditions for solid and solvent forms to preserve stability.
Catalog No. 50-002-97375 Supplier Medchemexpress LLC Supplier No. HYB0743A10MG
Pipecuronium bromide is a long-acting, nondepolarizing steroidal neuromuscular blocking agent supplied as a solid for research use. It functions as a nicotinic acetylcholine receptor antagonist to produce controlled muscle relaxation, and is commonly used in pharmacology and physiology studies requiring reproducible neuromuscular blockade.
  • Long-acting nondepolarizing neuromuscular blocker suitable for research.
  • High reported purity (≈99.9%) for reproducible experimental results.
  • Solid form enables stable storage and accurate dosing.
  • Available in small mass quantities compatible with laboratory assays.
  • Documented storage conditions for solid and solvent forms to preserve stability.