Learn More
MSC Honeywell Pipeliner Fiberglass Welding Helmet, Shade 10, Ratchet, 1 ea
Catalog No. 5000508659
Encompass
This fiberglass welding helmet is designed as an industry standard for pipe welding, providing a compact shell that allows for access to restricted work areas. It is engineered to withstand harsh welding conditions while ensuring user protection.
  • Compact design for use in tight workspaces
  • Fiberglass construction for durability
  • Ratchet suspension with constant-fit rubber headband for adjustable comfort
  • Fixed front mounting
  • Passive filter lens with shade 10
  • Viewing area of 2 in. x 4-1/4 in.
  • Impact resistant protection
  • Meets ANSI specifications
Catalog No. 50-005-08659 Supplier MSC Supplier No. 32362121
This fiberglass welding helmet is designed as an industry standard for pipe welding, providing a compact shell that allows for access to restricted work areas. It is engineered to withstand harsh welding conditions while ensuring user protection.
  • Compact design for use in tight workspaces
  • Fiberglass construction for durability
  • Ratchet suspension with constant-fit rubber headband for adjustable comfort
  • Fixed front mounting
  • Passive filter lens with shade 10
  • Viewing area of 2 in. x 4-1/4 in.
  • Impact resistant protection
  • Meets ANSI specifications