Learn More
Medchemexpress LLC Pipemidic acid trihydrate | 72571-82-5 | 98.0% | 357.36 | 100 MG
SDP
Catalog No. 5000454202
Encompass_Preferred
Pipemidic acid trihydrate is a derivative of Piromidic acid and functions as an antibacterial agent. It inhibits DNA gyrase. This compound is effective against gram-negative bacteria, including Pseudomonas aeruginosa, and also some gram-positive bacteria. It is suitable for research concerning intestinal, urinary, and biliary tract infections.
  • Inhibits DNA gyrase
  • Active against gram-negative bacteria, including Pseudomonas aeruginosa
  • Active against some gram-positive bacteria
  • Can be used for research of intestinal, urinary, and biliary tract infections
Catalog No. 50-004-54202 Supplier Medchemexpress LLC Supplier No. HYW013766100MG
Pipemidic acid trihydrate is a derivative of Piromidic acid and functions as an antibacterial agent. It inhibits DNA gyrase. This compound is effective against gram-negative bacteria, including Pseudomonas aeruginosa, and also some gram-positive bacteria. It is suitable for research concerning intestinal, urinary, and biliary tract infections.
  • Inhibits DNA gyrase
  • Active against gram-negative bacteria, including Pseudomonas aeruginosa
  • Active against some gram-positive bacteria
  • Can be used for research of intestinal, urinary, and biliary tract infections