Learn More
Medchemexpress LLC Pipemidic acid trihydrate | 72571-82-5 | 98.0% | 357.36 | 50 MG
SDP
Catalog No. 5000454589
Encompass_Preferred
Pipemidic acid trihydrate, a derivative of Piromidic acid, is an antibacterial agent that inhibits DNA gyrase. It is active against gram-negative bacteria, including Pseudomonas aeruginosa, as well as some gram-positive bacteria. This compound can be used for the research of intestinal, urinary, and biliary tract infections.
  • Derivative of Piromidic acid
  • Antibacterial agent
  • Inhibits DNA gyrase
  • Active against gram-negative bacteria, including Pseudomonas aeruginosa
  • Active against some gram-positive bacteria
  • Can be used for research of intestinal, urinary, and biliary tract infections
Catalog No. 50-004-54589 Supplier Medchemexpress LLC Supplier No. HYW01376650MG
Pipemidic acid trihydrate, a derivative of Piromidic acid, is an antibacterial agent that inhibits DNA gyrase. It is active against gram-negative bacteria, including Pseudomonas aeruginosa, as well as some gram-positive bacteria. This compound can be used for the research of intestinal, urinary, and biliary tract infections.
  • Derivative of Piromidic acid
  • Antibacterial agent
  • Inhibits DNA gyrase
  • Active against gram-negative bacteria, including Pseudomonas aeruginosa
  • Active against some gram-positive bacteria
  • Can be used for research of intestinal, urinary, and biliary tract infections