Learn More
Cayman Chemical Pipemidic Acid (hydrate), Purity: ≥98%, CAS: 72571-82-5, 25 g
SDP
Catalog No. 502946401
Encompass_Preferred
An antibiotic; active against clinical isolates of E. coli, P. mirabilis, P. inconstans, Shigella, Salmonella, Alcaligenes, and V. parahaemolyticus (MICs = 0.78-100 µg/ml), as well as drug-resistant clinical isolates of E. coli, P. mirabilis, Klebsiella, and Shigella (MICs = 1.56-6.25 µg/ml); Protective against systemic S. aureus, E. coli, K. pneumoniae, and P. aeruginosa infections in mice (ED50s = 237.5, 204.1, 28.6, and 99.5 mg/kg, respectively); protective against P. aeruginosa-induced pulmonary and dermal infections (ED50s = 81.7 and 173.2 mg/kg, respectively), as well as E. coli, K. pneumoniae, and P. aeruginosa urinary bladder infections (ED50s = 4.8, 11.9, and 30.6 mg/kg, respectively), in mice
Catalog No. 50-294-6401 Supplier Cayman Chemical Supplier No. 3371925 G
An antibiotic; active against clinical isolates of E. coli, P. mirabilis, P. inconstans, Shigella, Salmonella, Alcaligenes, and V. parahaemolyticus (MICs = 0.78-100 µg/ml), as well as drug-resistant clinical isolates of E. coli, P. mirabilis, Klebsiella, and Shigella (MICs = 1.56-6.25 µg/ml); Protective against systemic S. aureus, E. coli, K. pneumoniae, and P. aeruginosa infections in mice (ED50s = 237.5, 204.1, 28.6, and 99.5 mg/kg, respectively); protective against P. aeruginosa-induced pulmonary and dermal infections (ED50s = 81.7 and 173.2 mg/kg, respectively), as well as E. coli, K. pneumoniae, and P. aeruginosa urinary bladder infections (ED50s = 4.8, 11.9, and 30.6 mg/kg, respectively), in mice