Learn More
Medchemexpress LLC HY-13724B 5mg Medchemexpress, Pipendoxifene hydrochloride CAS:245124-69-0 Purity:>98%
SDP
Catalog No. 501932682
Encompass_Preferred
Medchemexpress, HY-13724B 5mg Pipendoxifene hydrochloride CAS:245124-69-0 Pipendoxifene hydrochloride is a selective estrogen receptor modulators (SERMs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2682 Supplier Medchemexpress LLC Supplier No. HY13724B5MG
Medchemexpress, HY-13724B 5mg Pipendoxifene hydrochloride CAS:245124-69-0 Pipendoxifene hydrochloride is a selective estrogen receptor modulators (SERMs). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.