Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Pipendoxifene hydrochloride | 245124-69-0 | 99.8% | 493.04 | 25 MG
SDP

Catalog No. 5000350973
Encompass_Preferred

Pipendoxifene hydrochloride is a selective estrogen receptor modulator (SERM) intended for breast cancer research. This 2-phenyl indole SERM demonstrates an excellent preclinical pharmacological profile, making it a candidate for further development as a treatment for metastatic breast cancer.

  • Purity: 99.83%
  • Appearance: Solid
  • Color: White to yellow
  • Target: Estrogen receptor
  • Molecular weight: 493.04
  • Solubility in DMSO: 100 mg/mL
  • Involved in a clinical trial for breast cancer (NCT00006369)
  • Recommended storage: 4°C, sealed, away from moisture; in solvent: -80°C for 6 months or -20°C for 1 month

Catalog No. 50-003-50973 Supplier Medchemexpress LLC Supplier No. 25MGHY13724B
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Pipendoxifene hydrochloride is a selective estrogen receptor modulator (SERM) intended for breast cancer research. This 2-phenyl indole SERM demonstrates an excellent preclinical pharmacological profile, making it a candidate for further development as a treatment for metastatic breast cancer.

  • Purity: 99.83%
  • Appearance: Solid
  • Color: White to yellow
  • Target: Estrogen receptor
  • Molecular weight: 493.04
  • Solubility in DMSO: 100 mg/mL
  • Involved in a clinical trial for breast cancer (NCT00006369)
  • Recommended storage: 4°C, sealed, away from moisture; in solvent: -80°C for 6 months or -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.