Learn More
Medchemexpress LLC Pipendoxifene hydrochloride | 245124-69-0 | 99.8% | C29H33ClN2O3 | 10MG
SDP
Catalog No. 5000260989
Encompass_Preferred
Pipendoxifene hydrochloride is a selective estrogen receptor modulator (SERM) used in preclinical research, including studies of breast cancer. Supplied as the hydrochloride salt, it is a white to yellow solid with high measured purity and recommended sealed storage to maintain stability. The compound is intended for research use and is provided with supporting safety documentation.
  • Selective estrogen receptor modulator for ER-targeted studies.
  • High purity suitable for research applications.
  • White to yellow solid form for convenient handling.
  • Multiple small-scale pack sizes available for screening.
  • Stable when stored sealed at recommended temperatures.
  • Provided with safety data sheet and technical information.
Catalog No. 50-002-60989 Supplier Medchemexpress LLC Supplier No. HY13724B10MG
Pipendoxifene hydrochloride is a selective estrogen receptor modulator (SERM) used in preclinical research, including studies of breast cancer. Supplied as the hydrochloride salt, it is a white to yellow solid with high measured purity and recommended sealed storage to maintain stability. The compound is intended for research use and is provided with supporting safety documentation.
  • Selective estrogen receptor modulator for ER-targeted studies.
  • High purity suitable for research applications.
  • White to yellow solid form for convenient handling.
  • Multiple small-scale pack sizes available for screening.
  • Stable when stored sealed at recommended temperatures.
  • Provided with safety data sheet and technical information.