Learn More
Medchemexpress LLC Pipendoxifene hydrochloride | 245124-69-0 | 99.83% | 493.04 | 1 ML
SDP
Catalog No. 5000348568
Encompass_Preferred
Pipendoxifene hydrochloride is a selective estrogen receptor modulator (SERM) developed for breast cancer research. It is a 2-phenyl indole SERM that has demonstrated an excellent preclinical pharmacological profile and has been selected for further development as a treatment for metastatic breast cancer.
  • Selective estrogen receptor modulator (SERM).
  • Used in research for breast cancer.
  • New 2-phenyl indole SERM with excellent preclinical pharmacological profile.
  • Available in various forms including solid and solution (e.g., 10 mM in DMSO).
  • High purity (99.83%).
Catalog No. 50-003-48568 Supplier Medchemexpress LLC Supplier No. HY13724B10MM/1ML
Pipendoxifene hydrochloride is a selective estrogen receptor modulator (SERM) developed for breast cancer research. It is a 2-phenyl indole SERM that has demonstrated an excellent preclinical pharmacological profile and has been selected for further development as a treatment for metastatic breast cancer.
  • Selective estrogen receptor modulator (SERM).
  • Used in research for breast cancer.
  • New 2-phenyl indole SERM with excellent preclinical pharmacological profile.
  • Available in various forms including solid and solution (e.g., 10 mM in DMSO).
  • High purity (99.83%).