Learn More
Medchemexpress LLC HY-100140 5mg Medchemexpress, Pipequaline CAS:77472-98-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1001405MG
Encompass_Preferred
Medchemexpress, HY-100140 5mg Pipequaline CAS:77472-98-1 Pipequaline (PK 8165) is a partial benzodiazepine receptor agonist with anxiolytic activity [1] [2] . Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1730