Learn More
Cayman Chemical Piperacetazine, Purity: ≥98%, CAS: 3819-00-9, 100 mg
SDP
Catalog No. 502910368
Encompass_Preferred
A typical antipsychotic; inhibits spontaneous motor activity and responding for food in rats, as well as potentiates hexobarbital-induced narcosis in mice; inhibits lysosome acidification, as well as viral entry (IC50 = <8.5 µM), in Ebola-infected Huh7 cells
Catalog No. 50-291-0368 Supplier Cayman Chemical Supplier No. 35220100 MG
A typical antipsychotic; inhibits spontaneous motor activity and responding for food in rats, as well as potentiates hexobarbital-induced narcosis in mice; inhibits lysosome acidification, as well as viral entry (IC50 = <8.5 µM), in Ebola-infected Huh7 cells